penta-Natriumtriphosphat ≥98,0% (durch titrimetrische Analyse), Purum p.a., Sigma-Aldrich®

Lieferant: Merck

Synonyme: Natriumtriphosphat pentabasisch, Natriumtriphosphat

M-Clarity™ Qualitätsstufe = MQ200

72061-500G
SIAL72061-500GEA 215 EUR
SIAL72061-500G
penta-Natriumtriphosphat ≥98,0% (durch titrimetrische Analyse), Purum p.a., Sigma-Aldrich®
penta-Natriumtriphosphat

For the determination of BSB acc. to DIN 38409-H 51.

Formel: Na₅O₁₀P₃
Molecular Weight: 367,86 g/mol
MDL: MFCD00003514
CAS-Nummer: 7758-29-4
EINECS: 231-838-7

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Colorless or White
Appearance (Form) Powder
Titration (Ion Exchange) 57-59 % calc. based on P₂O₅
Titration ≥ 98.0 %
Solubility (Color) Colorless
Solubility (Turbidity) Clear
Solubility (Method) 0.1 g IN 10 ml Water
pH 9.0 - 10.0
Metal Trace Analysis (ICP) Corresponds to Requirements
Calcium (ICP) ≤ 200 mg/kg
Cadmium (ICP) ≤ 50 mg/kg
Cobalt (ICP) ≤ 50 mg/kg
Copper (ICP) ≤ 50 mg/kg
Iron (ICP) ≤ 50 mg/kg
Nickel (ICP) ≤ 50 mg/kg
Lead (ICP) ≤ 50 mg/kg
Zinc (ICP) ≤ 50 mg/kg
Total Sulfur As SO₄ (ICP) ≤ 250 mg/kg
Chloride (Cl) ≤ 1000 mg/k

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR