2,3-Furandicarbonsäure, Sigma-Aldrich®

Lieferant: Merck

720658-5G
SIAL720658-5GEA 189 EUR
SIAL720658-5G
2,3-Furandicarbonsäure, Sigma-Aldrich®
Furan-2,3-dicarboxylic acid
Formel: C₆H₄O₅
Molecular Weight: 156,0930 g/mol
MDL: MFCD01860231
CAS-Nummer: 4282-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR