Ethyl-5-oxazolcarboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl oxazole-5-carboxylate

720763-1G
SIAL720763-1GEA 33.3 EUR
SIAL720763-1G
Ethyl-5-oxazolcarboxylat, Sigma-Aldrich®
Ethyl-5-oxazolcarboxylat
Formel: C₆H₇NO₃
Molecular Weight: 141,13 g/mol
MDL: MFCD04114930
CAS-Nummer: 118994-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR