1H-Indazol-6-boronsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1H-Indazol-6-ylboronic acid, 6-Indazolyboronic acid, 1H-Indazol-6-yl-6-boronic acid
720828-250MG
SIAL720828-250MGEA
156
EUR
SIAL720828-250MG
1H-Indazol-6-boronsäure, Sigma-Aldrich®
1H-Indazole-6-boronic acid
Formel:
C₇H₇BN₂O₂ Molecular Weight: 161,9500 g/mol |
MDL:
MFCD04972419 CAS-Nummer: 885068-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...