1-Iodisochinolin, Sigma-Aldrich®

Lieferant: Merck

720879-1G
SIAL720879-1GEA 89.4 EUR
SIAL720879-1G
1-Iodisochinolin, Sigma-Aldrich®
1-Iodoisoquinoline
Formel: C₉H₆IN
Molecular Weight: 255,0551 g/mol
MDL: MFCD00234494
CAS-Nummer: 19658-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR