3-Amino-5-brom-2-chlorpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Brom-2-chlor-3-pyridylamin

720909-5G
SIAL720909-5GEA 53.8 EUR
SIAL720909-5G
3-Amino-5-brom-2-chlorpyridin, Sigma-Aldrich®
3-Amino-5-bromo-2-chloropyridine
Formel: C₅H₄BrClN₂
Molecular Weight: 207,46 g/mol
MDL: MFCD02682092
CAS-Nummer: 588729-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR