3-Carboxy-5-nitrophenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Carboxy-5-nitrobenzeneboronic acid, 3-Borono-5-nitrobenzoic acid

721042-5G
SIAL721042-5GEA 122 EUR
SIAL721042-5G
3-Carboxy-5-nitrophenylboronsäure, Sigma-Aldrich®
3-Carboxy-5-nitrophenylboronsäure
Formel: C₇H₆BNO₆
Molecular Weight: 210.94 g/mol
MDL: MFCD00757433
CAS-Nummer: 101084-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR