2-Methyl-6-(tributylstannyl)pyridin, Sigma-Aldrich®

Lieferant: Merck
Gefahr

721220-1G
SIAL721220-1GEA 263 EUR
SIAL721220-1G
2-Methyl-6-(tributylstannyl)pyridin, Sigma-Aldrich®
2-Methyl-6-(tributylstannyl)pyridin
Formel: C₁₈H₃₃NSn
Molecular Weight: 382,16 g/mol
MDL: MFCD07787401
CAS-Nummer: 259807-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR