2,5-Furandicarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,5-Furandicarboxylic acid

722081-5G
SIAL722081-5GEA 242 EUR
SIAL722081-5G
2,5-Furandicarbonsäure, Sigma-Aldrich®
Furan-2,5-dicarboxylic acid
Formel: C₆H₄O₅
Molecular Weight: 156,09 g/mol
MDL: MFCD00016582
CAS-Nummer: 3238-40-2
EINECS: 221-800-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR