α-Brom-2-chlorphenylessigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Bromo-2-(2-chlorophenyl)acetic acid

722235-25G
SIAL722235-25GEA 38.4 EUR
SIAL722235-25G
α-Brom-2-chlorphenylessigsäure, Sigma-Aldrich®
α-Brom-2-chlorphenylessigsäure
Formel: C₈H₆BrClO₂
Molecular Weight: 249,49 g/mol
MDL: MFCD03428513
CAS-Nummer: 29270-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR