3-Iod-4-methoxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

722278-1G
SIAL722278-1GEA 42.3 EUR
SIAL722278-1G
3-Iod-4-methoxybenzaldehyd, Sigma-Aldrich®
3-Iodo-4-methoxybenzaldehyde
Formel: C₈H₇IO₂
Molecular Weight: 262,04 g/mol
MDL: MFCD01009500
CAS-Nummer: 2314-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR