2-Methoxy-4-methyl-5-nitropyridin, Sigma-Aldrich®

Lieferant: Merck

722286-1G
SIAL722286-1GEA 83.7 EUR
SIAL722286-1G
2-Methoxy-4-methyl-5-nitropyridin, Sigma-Aldrich®
2-Methoxy-4-methyl-5-nitropyridine
Formel: C₇H₈N₂O₃
Molecular Weight: 168,15 g/mol
MDL: MFCD03095075
CAS-Nummer: 6635-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR