6-Hydroxy-m-anisidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-4-methoxyphenol

722367-1G
SIAL722367-1GEA 48.6 EUR
SIAL722367-1G
6-Hydroxy-m-anisidin, Sigma-Aldrich®
6-Hydroxy-m-anisidin
Formel: C₇H₉NO₂
Molecular Weight: 139,16 g/mol
MDL: MFCD06616911
CAS-Nummer: 20734-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR