3-Amino-5-brom-2-fluorpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromo-2-fluoropyridin-3-amine

722391-5G
SIAL722391-5GEA 78.8 EUR
SIAL722391-5G
3-Amino-5-brom-2-fluorpyridin, Sigma-Aldrich®
3-Amino-5-bromo-2-fluoropyridine
Formel: C₅H₄BrFN₂
Molecular Weight: 191.0011 g/mol
MDL: MFCD06659524
CAS-Nummer: 884495-22-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR