2-Brom-4-chlorpyridin, Sigma-Aldrich®

Lieferant: Merck

722413-5G
SIAL722413-5GEA 123 EUR
SIAL722413-5G
2-Brom-4-chlorpyridin, Sigma-Aldrich®
2-Bromo-4-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192.44 g/mol
MDL: MFCD03840757
CAS-Nummer: 22918-01-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR