5-Brom-o-kresol, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 5-Brom-2-methylphenol, 4-Brom-2-hydroxytoluol, 1-Brom-3-hydroxy-4-methylbenzol

722480-10G
SIAL722480-10GEA 105 EUR
SIAL722480-10G
5-Brom-o-kresol, Sigma-Aldrich®
5-Brom-o-kresol
Formel: C₇H₇BrO
Molecular Weight: 187.04 g/mol
MDL: MFCD08061906
CAS-Nummer: 36138-76-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR