5-Isochinolincarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Isoquinoline-5-carboxylic acid

722626-250MG
SIAL722626-250MGEA 35.4 EUR
SIAL722626-250MG
5-Isochinolincarbonsäure, Sigma-Aldrich®
5-Isoquinolinecarboxylic acid
Formel: C₁₀H₇NO₂
Molecular Weight: 173,17 g/mol
MDL: MFCD03788744
CAS-Nummer: 27810-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR