Tris(2-pyridylmethyl)amine 98% (durch HPLC), Sigma-Aldrich®

Lieferant: Merck
Achtung

723134-250MG
SIAL723134-250MGEA 0 EUR
SIAL723134-250MG
Tris(2-pyridylmethyl)amine 98% (durch HPLC), Sigma-Aldrich®
Tris(2-pyridylmethyl)amine
Formel: C₁₈H₁₈N₄
Molecular Weight: 290,37 g/mol
Schmelzpunkt: 85,..89 °C
MDL: MFCD14708176
CAS-Nummer: 16858-01-8

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Dark Yellow and Faint Beige to Dark Beige
Appearance (Form) Conforms to Requirements
Proton NMR Spectrum Conforms to Structure
Carbon 72.6 - 76.3%
Nitrogen 18.8 - 19.8%
Purity (HPLC) ≥97.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR