2-Brom-4-fluorpyridin, Sigma-Aldrich®

Lieferant: Merck

723339-1G
SIAL723339-1GEA 61.7 EUR
SIAL723339-1G
2-Brom-4-fluorpyridin, Sigma-Aldrich®
2-Bromo-4-fluoropyridine
Formel: C₅H₃BrFN
Molecular Weight: 175,99 g/mol
MDL: MFCD09260909
CAS-Nummer: 357927-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR