Kalium(2-nitrophenyl)trifluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-Nitrophenyl)trifluorborsäure Kaliumsalz

723592-1G
SIAL723592-1GEA 125 EUR
SIAL723592-1G
Kalium(2-nitrophenyl)trifluoroborat, Sigma-Aldrich®
Kalium(2-nitrophenyl)trifluoroborat
Formel: C₆H₄BF₃KNO₂
Molecular Weight: 229,01 g/mol
MDL: MFCD04115759
CAS-Nummer: 850623-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR