4-(N,N-Diphenylamino)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Diphenylamino)phenylboronic acid Pinacol ester, 4-(Diphenylamino)benzeneboronic acid pinacol ester, Triphenylamine-4-boronic acid Pinacol ester, N,N-Diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-[4-(Diphenylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

723614-1G 723614-5G
SIAL723614-1GEA 74 EUR
SIAL723614-1G SIAL723614-5G
4-(N,N-Diphenylamino)phenylboronsäurepinakolester, Sigma-Aldrich®
4-(N,N-Diphenylamino)phenylboronsäurepinakolester
Formel: C₂₄H₂₆BNO₂
MDL: MFCD13195770
CAS-Nummer: 267221-88-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR