3-Ethyl-1-pentin-3-ol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Ethyl-3-hydroxy-1-pentin, 3-Ethylpent-1-in-3-ol

723800-5G
SIAL723800-5GEA 166 EUR
SIAL723800-5G
3-Ethyl-1-pentin-3-ol, Sigma-Aldrich®
3-Ethyl-1-pentin-3-ol
Formel: C₇H₁₂O
Molecular Weight: 112.17 g/mol
MDL: MFCD00014392
CAS-Nummer: 6285-06-9
EINECS: 228-512-1
UN: 1987
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR