2,3-Dichlor-5-nitropyridin, Sigma-Aldrich®

Lieferant: Merck

724297-1G
SIAL724297-1GEA 57.3 EUR
SIAL724297-1G
2,3-Dichlor-5-nitropyridin, Sigma-Aldrich®
2,3-Dichloro-5-nitropyridine
Formel: C₅H₂Cl₂N₂O₂
MDL: MFCD03840432
CAS-Nummer: 22353-40-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR