Methyl-4-ethinylbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 4-ethynylbenzoate

724653-1G
SIAL724653-1GEA 67.8 EUR
SIAL724653-1G
Methyl-4-ethinylbenzoat, Sigma-Aldrich®
Methyl-4-ethinylbenzoat
Formel: C₁₀H₈O₂
Molecular Weight: 160.17 g/mol
MDL: MFCD00168820
CAS-Nummer: 3034-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR