3-Methoxy-p-toluidin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Methyl-m-anisidin, 5-Amino-2-methylanisol, 3-Methoxy-4-methylanilin, 4-Amino-2-methoxytoluol, 1-Amino-3-methoxy-4-methylbenzol
724688-1G
SIAL724688-1GEA
103
EUR
SIAL724688-1G
3-Methoxy-p-toluidin, Sigma-Aldrich®
3-Methoxy-p-toluidin
Formel:
C₈H₁₁NO Molecular Weight: 137,18 g/mol Siedepunkt: 250 °C (1013 hPa) Schmelzpunkt: 58 °C |
MDL:
MFCD00025371 CAS-Nummer: 16452-01-0 EINECS: 240-500-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...