3-Aminobenzolsulfonamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Anilin-3-sulfonsäureamid, 3-Aminobenzolsulfonsäureamid

724696-5G
SIAL724696-5GEA 195 EUR
SIAL724696-5G
3-Aminobenzolsulfonamid, Sigma-Aldrich®
3-Aminobenzolsulfonamid
Formel: C₆H₈N₂O₂S
Molecular Weight: 172,21 g/mol
MDL: MFCD00035781
CAS-Nummer: 98-18-0
EINECS: 202-646-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR