3-Amino-2-brombenzoesäure, Sigma-Aldrich®

Lieferant: Merck

724920-500MG
SIAL724920-500MGEA 94.7 EUR
SIAL724920-500MG
3-Amino-2-brombenzoesäure, Sigma-Aldrich®
3-Amino-2-bromobenzoic acid
Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
MDL: MFCD07786957
CAS-Nummer: 168899-61-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR