(2-Brom-4-pyridinyl)methanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Bromopyridine-4-methanol, 2-Bromo-4-pyridinemethanol

725072-1G
SIAL725072-1GEA 55.6 EUR
SIAL725072-1G
(2-Brom-4-pyridinyl)methanol, Sigma-Aldrich®
(2-Brom-4-pyridinyl)methanol
Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
MDL: MFCD04039313
CAS-Nummer: 118289-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR