6-Sulfanyl-1-hexanol, Sigma-Aldrich®

Lieferant: Merck

725226-1G
SIAL725226-1GEA 278 EUR
SIAL725226-1G
6-Sulfanyl-1-hexanol, Sigma-Aldrich®
6-Mercapto-1-hexanol
Formel: C₆H₁₄OS
Molecular Weight: 134,24 g/mol
MDL: MFCD00068552
CAS-Nummer: 1633-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR