Rhodium(II) triphenylacetate dimer (CH₂Cl₂ complex), Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Rhodium(II) 2,2,2-triphenylacetate dimer, Tetrakis(triphenylacetato)dirhodium(II) dichloromethane, Rh2(triphenylacetate)4, Rh2(TPA)4, Tetrakis(triphenylacetato)dirhodium(II), Rhodiumtriphenylacetate, Rhodium(II) Bis(triphenylacetate) dimer dichloromethane adduct, Rh2(TPA)4 dichloromethane adduct, Dirhodium(II) Tetrakis(triphenylacetate) dichloromethane adduct, Rhodium triphenyl acetate

M-Clarity™ Qualitätsstufe = MQ100

725455-100MG
SIAL725455-100MGEA 129 EUR
SIAL725455-100MG
Rhodium(II) triphenylacetate dimer (CH₂Cl₂ complex), Sigma-Aldrich®
Rhodium(II) triphenylacetate dimer
Formel: C₈₀H₆₀O₈Rh₂·CH₂Cl₂
Schmelzpunkt: 260-263 °C
MDL: MFCD15071082
CAS-Nummer: 142214-04-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR