Hafniumtetrakis[ethyl(methyl)azanid], Sigma-Aldrich®

Lieferant: Merck

725544-10G
SIAL725544-10GEA 1270 EUR
SIAL725544-10G
Hafniumtetrakis[ethyl(methyl)azanid], Sigma-Aldrich®
Tetrakis(ethylmethylamino)hafnium(IV)
Formel: C₁₂H₃₂HfN₄
MDL: MFCD03427130
CAS-Nummer: 352535-01-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR