(R)-(+)-Epoxystyrol, Sigma-Aldrich®

Lieferant: Merck

726508-25G
SIAL726508-25GEA 427 EUR
SIAL726508-25G
(R)-(+)-Epoxystyrol, Sigma-Aldrich®
(R)-(+)-Epoxystyrol
Formel: C₈H₈O
MDL: MFCD00066210
CAS-Nummer: 20780-53-4
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR