(R)-(+)-4-Fluor-α-methylbenzylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-(+)-1-(4-Fluorophenyl)ethylamine, (R)-1-(4-Fluorophenyl)ethanamine

727024-5G
SIAL727024-5GEA 0 EUR
SIAL727024-5G
(R)-(+)-4-Fluor-α-methylbenzylamin, Sigma-Aldrich®
(R)-(+)-4-Fluor-α-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139.17 g/mol
MDL: MFCD03093090
CAS-Nummer: 374898-01-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR