3-Bromdiphenylether (BDE Nr. 2), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Bromodiphenyl ether, 1-Bromo-3-phenoxybenzene

728969-1G
SIAL728969-1GEA 63.6 EUR
SIAL728969-1G
3-Bromdiphenylether (BDE Nr. 2), Sigma-Aldrich®
3-Bromdiphenylether (BDE Nr. 2)
Formel: C₁₂H₉BrO
Molecular Weight: 249,1000 g/mol
MDL: MFCD00474520
CAS-Nummer: 6876-00-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR