4-Chlor-2,6-difluorbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

728985-1G
SIAL728985-1GEA 47.1 EUR
SIAL728985-1G
4-Chlor-2,6-difluorbenzaldehyd, Sigma-Aldrich®
4-Chloro-2,6-difluorobenzaldehyde
Formel: C₇H₃ClF₂O
Molecular Weight: 176,55 g/mol
MDL: MFCD03094460
CAS-Nummer: 252004-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR