2-Brom-6-chlorpyridin, Sigma-Aldrich®

Lieferant: Merck

729388-1G
SIAL729388-1GEA 66.7 EUR
SIAL729388-1G
2-Brom-6-chlorpyridin, Sigma-Aldrich®
2-Bromo-6-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192.44 g/mol
MDL: MFCD00181262
CAS-Nummer: 5140-72-7
EINECS: 225-904-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR