Ethyl-4-nitrobenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Nitrobenzoesäureethylester

72940-100G
SIAL72940-100GEA 53 EUR
SIAL72940-100G
Ethyl-4-nitrobenzoat, Sigma-Aldrich®
Ethyl-4-nitrobenzoat
Formel: C₉H₉NO₄
Molecular Weight: 195,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007351
CAS-Nummer: 99-77-4
EINECS: 202-786-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR