Methyl-1H-indazol-4-carboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 1H-indazole-4-carboxylate

729744-1G
SIAL729744-1GEA 84.3 EUR
SIAL729744-1G
Methyl-1H-indazol-4-carboxylat, Sigma-Aldrich®
Methyl-1H-indazol-4-carboxylat
Formel: C₉H₈N₂O₂
Molecular Weight: 176,1700 g/mol
MDL: MFCD07371610
CAS-Nummer: 192945-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR