2-(Trifluormethyl)pyrazin, Sigma-Aldrich®

Lieferant: Merck

730025-1G
SIAL730025-1GEA 83.1 EUR
SIAL730025-1G
2-(Trifluormethyl)pyrazin, Sigma-Aldrich®
2-(Trifluoromethyl)pyrazine
Formel: C₅H₃F₃N₂
Molecular Weight: 148,09 g/mol
MDL: MFCD06797734
CAS-Nummer: 61655-67-2
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR