2,3-Dichlor-4-iodpyridin, Sigma-Aldrich®

Lieferant: Merck

730564-1G
SIAL730564-1GEA 81.1 EUR
SIAL730564-1G
2,3-Dichlor-4-iodpyridin, Sigma-Aldrich®
2,3-Dichloro-4-iodopyridine
Formel: C₅H₂Cl₂IN
Molecular Weight: 273,8900 g/mol
MDL: MFCD11857737
CAS-Nummer: 889865-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR