2-Iod-3-nitrotoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Iodo-1-methyl-3-nitrobenzene

730742-5G
SIAL730742-5GEA 61.3 EUR
SIAL730742-5G
2-Iod-3-nitrotoluol, Sigma-Aldrich®
2-Iod-3-nitrotoluol
Formel: C₇H₆INO₂
Molecular Weight: 263,0300 g/mol
MDL: MFCD00091040
CAS-Nummer: 6277-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR