3-Fluor-o-anisidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Fluor-2-methoxyanilin, 2-Amino-6-fluoranisol, 3-Fluor-2-anisidin

731080-1G
SIAL731080-1GEA 28.5 EUR
SIAL731080-1G
3-Fluor-o-anisidin, Sigma-Aldrich®
3-Fluor-o-anisidin
Formel: C₇H₈FNO
Molecular Weight: 141,15 g/mol
MDL: MFCD00007759
CAS-Nummer: 437-83-2
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR