8-Bromo-3-methoxyisoquinoline, Sigma-Aldrich®

Lieferant: Merck

731358-1G
SIAL731358-1GEA 103 EUR
SIAL731358-1G
8-Bromo-3-methoxyisoquinoline, Sigma-Aldrich®
8-Bromo-3-methoxyisoquinoline
Formel: C₁₀H₈BrNO
Molecular Weight: 238,08 g/mol
CAS-Nummer: 608515-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR