2-Fluor-3-iod-5-methylpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluor-3-iod-5-picolin

731390-250MG
SIAL731390-250MGEA 22.1 EUR
SIAL731390-250MG
2-Fluor-3-iod-5-methylpyridin, Sigma-Aldrich®
2-Fluoro-3-iodo-5-methylpyridine
Formel: C₆H₅FIN
Molecular Weight: 237,02 g/mol
MDL: MFCD03095303
CAS-Nummer: 153034-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR