2-Fluor-4-iod-5-methylpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluor-4-iod-5-picolin

731404-250MG
SIAL731404-250MGEA 29.5 EUR
SIAL731404-250MG
2-Fluor-4-iod-5-methylpyridin, Sigma-Aldrich®
2-Fluoro-4-iodo-5-methylpyridine
Formel: C₆H₅FIN
Molecular Weight: 237,02 g/mol
MDL: MFCD03095297
CAS-Nummer: 153034-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR