4,4′,4″-s-Triazine-2,4,6-triyl-tribenzoic acid ≥95%, Sigma-Aldrich®

Lieferant: Merck
Achtung

741175-500MG
SIAL741175-500MGEA 182 EUR
SIAL741175-500MG
4,4′,4″-s-Triazine-2,4,6-triyl-tribenzoic acid ≥95%, Sigma-Aldrich®
4,4′,4″-s-Triazine-2,4,6-triyl-tribenzoic acid

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency.

Formel: C₂₄H₁₅N₃O₆
Schmelzpunkt: 83…88 °C
MDL: MFCD04116314
CAS-Nummer: 61414-16-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR