Tris[3,5-bis(trifluormethyl)phenyl]phosphin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tris(3,5-bis(trifluoromethyl)phenyl)phosphine

74231-500MG-F
SIAL74231-500MG-FEA 233 EUR
SIAL74231-500MG-F
Tris[3,5-bis(trifluormethyl)phenyl]phosphin, Sigma-Aldrich®
Tris[3,5-bis(trifluormethyl)phenyl]phosphin
Formel: C₂₄H₉F₁₈P
Molecular Weight: 670,28 g/mol
MDL: MFCD00042428
CAS-Nummer: 175136-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR