DNA (Deoxyribonucleic acid) Sperm_Fish, Sigma-Aldrich®

Lieferant: Merck


MQ100

74782-100G 74782-10G
SIAL74782-100GEA 415 EUR
SIAL74782-100G SIAL74782-10G
DNA (Deoxyribonucleic acid) Sperm_Fish, Sigma-Aldrich®
DNS (Desoxyribonukleinsäure)
MDL: MFCD00283430
CAS-Nummer: 9007-49-2
EINECS: 309-566-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR