1-Butyl-3-methylimidazoliumoctylsulfat, Sigma-Aldrich®

Lieferant: Merck

75059-50G-F 75059-5G-F
SIAL75059-50G-FEA 359 EUR
SIAL75059-50G-F SIAL75059-5G-F
1-Butyl-3-methylimidazoliumoctylsulfat, Sigma-Aldrich®
1-Butyl-3-methylimidazolium octylsulphate
Formel: C₁₆H₃₂N₂O₄S
MDL: MFCD03788914
CAS-Nummer: 445473-58-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR