Olivenöl, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

75343-1L
SIAL75343-1LEA 116 EUR
SIAL75343-1L
Olivenöl, Sigma-Aldrich®
Olive oil
MDL: MFCD00131764
CAS-Nummer: 8001-25-0
EINECS: 232-277-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR